Learn More
System Biosciences LLC pCDH-MSCV-MCS-EF1α-Puro Cloning and Expression LentivectorpCDH-MSCV-MCS-EF1α-Puro cDNA Dual Promoter Cloning and Expression Lentivector = 10ug
SDP
Catalog No. NC2240558
Encompass_Preferred
The pCDH-MSCV-MCS-EF1α-Puro lentivector uses the MSCV promoter for high expression in hematopoietic or stem cells and includes a puromycin resistance marker. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-MSCV-MCS-EF1α-Puro Cloning and Expression Lentivector drives expression of your gene-of-interest from the MSCV promoter, which provides strong expression in hematopoietic and stem cells. This lentivector also enables selection for transductants using the puromycin resistance marker.
This item is not returnable. View return policy
Catalog No. NC2240558 Supplier System Biosciences LLC Supplier No. CD710B1
The pCDH-MSCV-MCS-EF1α-Puro lentivector uses the MSCV promoter for high expression in hematopoietic or stem cells and includes a puromycin resistance marker. Well-regarded in the industry for reliable gene expression, SBI’s lentiviral vectors come in a variety of formats that support a wide range of applications. The pCDH-MSCV-MCS-EF1α-Puro Cloning and Expression Lentivector drives expression of your gene-of-interest from the MSCV promoter, which provides strong expression in hematopoietic and stem cells. This lentivector also enables selection for transductants using the puromycin resistance marker.