Learn More
Aviva Systems Biology Corp Pcdh10 Antibody - C-terminal region (ARP41634_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41634_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pcdh10. It was validated on Western Blot. . Alternate names: 6430521D13Rik, 6430703F07Rik, OL-pc, mKIAA1400ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-948