Learn More
Aviva Systems Biology Corp PCDH12 Antibody - middle region (ARP47275_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47275_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDH12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCDH12. Alternate names: VE-cadherin-2, VECAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-53511