Learn More
Aviva Systems Biology Corp PCDH15 Antibody - N-terminal region (ARP50153_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50153_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDH15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDH15. Alternate names: RP11-449J3.2, DFNB23, DKFZp667A1711, USH1F, CDHR15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53620