Learn More
Aviva Systems Biology Corp PCDH17 Antibody - C-terminal region (ARP54517_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54517_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDH17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PCDH17. Alternate names: PCDH68, PCH68ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61230