Learn More
Aviva Systems Biology Corp PCDH18 Antibody - N-terminal region (ARP49430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDH18. It was validated on Western Blot. PCDH18,protocadherin 18,PCDH68L; KIAA1562; DKFZp434B0923; PCDH18,protocadherin-18,protocadherin-18,OTTHUMP00000220081,OTTHUMP00000220082,protocadherin 68-like proteinThe immunogen is a synthetic peptide directed towards the N terminal region of human PCDH18. Alternate names: DKFZP434B0923, KIAA1562, PCDH68LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-418