Learn More
Aviva Systems Biology Corp PCDHA11 Antibody - N-terminal region (ARP49346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHA11. It was validated on Western Blot. . Alternate names: CNR7, CNRN7, CNRS7, CRNR7, PCDH-ALPHA11ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-412