Learn More
Aviva Systems Biology Corp PCDHA6 Antibody - C-terminal region (ARP50008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PCDHA6. Alternate names: CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60238