Learn More
Aviva Systems Biology Corp PCDHB15 Antibody - N-terminal region (ARP49401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHB15. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHB15. Alternate names: PCDH-BETA15ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-285-413