Learn More
Aviva Systems Biology Corp PCDHB16 Antibody - middle region (ARP49598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHB16. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PCDHB16. Alternate names: KIAA1621, ME1, PCDH-BETA16, PCDH3X, PCDHB8aApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rat
Catalog No. 50-285-438