Learn More
Aviva Systems Biology Corp PCDHGB1 Antibody - N-terminal region (ARP49377_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49377_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHGB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHGB1. Alternate names: MGC119466, MGC119467, MGC119469, PCDH-GAMMA-B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60075