Learn More
Aviva Systems Biology Corp PCDHGC3 Antibody - N-terminal region (ARP45334_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45334_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHGC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHGC3. Alternate names: PC43, PCDH-GAMMA-C3, PCDH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-59003