Learn More
Aviva Systems Biology Corp PCDHGC4 Antibody - N-terminal region (ARP49389_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49389_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHGC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHGC4. Alternate names: MGC119489, PCDH-GAMMA-C4ApplicationWB, IHC. Species reactivity: Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53582