Learn More
Electron Microscopy Sciences Castro Viejo Needle Holder, PCF-TXCC
SDP
Catalog No. 5030634
Encompass_Preferred
Half Lunar Jaws Forceps have fine diamond dust inside jaws to reinforce gripping capability. When the jaws are fully open the diameter is 8mm and when closed it is 2mm.
Model: PCF-TXCC
OAL: 135 mm
Catalog No. 50-306-34 Supplier Electron Microscopy Sciences Supplier No. 77894S
Half Lunar Jaws Forceps have fine diamond dust inside jaws to reinforce gripping capability. When the jaws are fully open the diameter is 8mm and when closed it is 2mm.
Model: PCF-TXCC
OAL: 135 mm