Learn More
Aviva Systems Biology Corp PCGF1 Antibody - middle region (ARP43370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCGF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PCGF1. Alternate names: 2010002K04Rik, FLJ43754, MGC10882, NSPC1, RNF3A-2, RNF68ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-068