Learn More
Aviva Systems Biology Corp PCGF5 Antibody - N-terminal region (ARP43358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCGF5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCGF5. Alternate names: MGC16202, RNF159ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58506