Learn More
Apexbio Technology LLC PCI-24781 (CRA-024781) 783355-60-2 200mg
SDP
Catalog No. 5000062544
Encompass_Preferred
PCI-24781 (CRA-024781 CAS 783355-60-2) is a histone deacetylase (HDAC) inhibitor targeting HDAC1 HDAC2 HDAC3 HDAC6 HDAC8 and HDAC10 with reported Ki values of 7 19 8 2 17 280 and 24 nM respectively HDAC enzymes catalyze the removal of acetyl groups from lysine residues on histone proteins thereby regulating chromatin structure and gene expression PCI-24781 induces dose-dependent hyperacetylation of histones and tubulin in human cancer cells (HCT116 DLD-1) leading to apoptosis via p21 upregulation PARP cleavage and H2AX accumulation PCI-24781 demonstrates antitumor activity against multiple tumor cell lines and in xenograft models highlighting its utility for oncology research
Catalog No. 50-000-62544 Supplier Apexbio Technology LLC Supplier No. A4098200
PCI-24781 (CRA-024781 CAS 783355-60-2) is a histone deacetylase (HDAC) inhibitor targeting HDAC1 HDAC2 HDAC3 HDAC6 HDAC8 and HDAC10 with reported Ki values of 7 19 8 2 17 280 and 24 nM respectively HDAC enzymes catalyze the removal of acetyl groups from lysine residues on histone proteins thereby regulating chromatin structure and gene expression PCI-24781 induces dose-dependent hyperacetylation of histones and tubulin in human cancer cells (HCT116 DLD-1) leading to apoptosis via p21 upregulation PARP cleavage and H2AX accumulation PCI-24781 demonstrates antitumor activity against multiple tumor cell lines and in xenograft models highlighting its utility for oncology research