Learn More
Selleck Chemical LLC Abexinostat (PCI-24781) 50mg 783355-60-2 CRA-024781
SDP
Catalog No. 508413
Encompass_Preferred
Abexinostat (PCI-24781, CRA-024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM, modest potent to HDACs 2, 3, 6, and 10 and greater than 40-fold selectivity against HDAC8. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-841-3 Supplier Selleck Chemical LLC Supplier No. S109050MG
Abexinostat (PCI-24781, CRA-024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM, modest potent to HDACs 2, 3, 6, and 10 and greater than 40-fold selectivity against HDAC8. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.