Learn More
Apexbio Technology LLC PCI-32765 (Ibrutinib) 936563-96-1 5mg
SDP
Catalog No. 5000064217
Encompass_Preferred
PCI-32765 (Ibrutinib) is a small-molecule inhibitor targeting Bruton s tyrosine kinase (BTK) a cytoplasmic kinase integral to B-cell receptor (BCR) signaling By covalently binding to BTK active sites PCI-32765 disrupts downstream signaling cascades essential for B-cell maturation and activation Dysfunction or inhibition of BTK correlates with impaired B-cell development and reduction in autoantibody production PCI-32765 is commonly utilized in biomedical research to explore BTK-dependent signaling pathways elucidate molecular mechanisms underlying B-cell mediated disorders and examine therapeutic potential in B-cell malignancies and autoimmune models
Catalog No. 50-000-64217 Supplier Apexbio Technology LLC Supplier No. A30015
PCI-32765 (Ibrutinib) is a small-molecule inhibitor targeting Bruton s tyrosine kinase (BTK) a cytoplasmic kinase integral to B-cell receptor (BCR) signaling By covalently binding to BTK active sites PCI-32765 disrupts downstream signaling cascades essential for B-cell maturation and activation Dysfunction or inhibition of BTK correlates with impaired B-cell development and reduction in autoantibody production PCI-32765 is commonly utilized in biomedical research to explore BTK-dependent signaling pathways elucidate molecular mechanisms underlying B-cell mediated disorders and examine therapeutic potential in B-cell malignancies and autoimmune models