Learn More
Selleck Chemical LLC Ibrutinib (PCI-32765) 5mg 936563-96-1
SDP
Catalog No. NC0342942
Encompass_Preferred
Ibrutinib (PCI-32765) is a potent and highly selective Brutons tyrosine kinase (Btk) inhibitor with IC50 of 0.5 nM in cell-free assays, modestly potent to Bmx, CSK, FGR, BRK, HCK, less potent to EGFR, Yes, ErbB2, JAK3, etc. Ibrutinib is applicable as a Btk ligand in the synthesis of a series of PROTACs including P13I. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0342942 Supplier Selleck Chemical LLC Supplier No. S26805MG
Ibrutinib (PCI-32765) is a potent and highly selective Brutons tyrosine kinase (Btk) inhibitor with IC50 of 0.5 nM in cell-free assays, modestly potent to Bmx, CSK, FGR, BRK, HCK, less potent to EGFR, Yes, ErbB2, JAK3, etc. Ibrutinib is applicable as a Btk ligand in the synthesis of a series of PROTACs including P13I. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.