Learn More
Medchemexpress LLC HY-100335 50mg Medchemexpress, PCI-33380 CAS:1022899-36-0 Purity:>98%
SDP
Catalog No. 501931777
Encompass_Preferred
Medchemexpress, HY-100335 50mg PCI-33380 CAS:1022899-36-0 PCI-33380 is an irreversible Bruton's Tyrosine Kinase (BTK) inhibitor (fluorescent probe). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1777 Supplier Medchemexpress LLC Supplier No. HY10033550MG
Medchemexpress, HY-100335 50mg PCI-33380 CAS:1022899-36-0 PCI-33380 is an irreversible Bruton's Tyrosine Kinase (BTK) inhibitor (fluorescent probe). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.