Learn More
Selleck Chemical LLC PCI-34051 10mg 950762-95-5
SDP
Catalog No. NC0568024
Encompass_Preferred
PCI-34051 is a potent and specific HDAC8 inhibitor with IC50 of 10 nM in a cell-free assay. It has greater than 200-fold selectivity over HDAC1 and 6, more than 1000-fold selectivity over HDAC2, 3, and 10. PCI-34051 induces caspase-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0568024 Supplier Selleck Chemical LLC Supplier No. S201210MG
PCI-34051 is a potent and specific HDAC8 inhibitor with IC50 of 10 nM in a cell-free assay. It has greater than 200-fold selectivity over HDAC1 and 6, more than 1000-fold selectivity over HDAC2, 3, and 10. PCI-34051 induces caspase-dependent apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.