Learn More
Cayman Chemical PCI 34051, 950762-95-5, 5 mg
SDP
Supplier:  Cayman Chemical 104445
Encompass_Preferred
Molecular Formula C17H16N2O3, Purity ≥95%, Formula Weight 296.3; A potent HDAC8 inhibitor (IC50 = 0.01 M) with >200-fold selectivity over HDAC isoforms 1, 2, 3, 6, and 10 (IC50s = 4, >50, >50, 2.9, and 13 M, respectively); induces caspase-dependent apoptosis in cell lines derived from T-cell lymphomas or leukemias (GI50s = 2.4 - 4 M).
Catalog No. NC0588318