Learn More
New England Biolabs, Inc. PciI – 1000 units
SDP
Catalog No. 50994898
Encompass
A restriction endonuclease that recognizes the sequence A^CATG_T.
Catalog No. 50-994-898 Supplier New England Biolabs, Inc. Supplier No. R0655L
A restriction endonuclease that recognizes the sequence A^CATG_T.