Learn More
New England Biolabs, Inc. PciI – 200 units
SDP
Catalog No. 50812683
Encompass
A restriction endonuclease that recognizes the sequence A^CATG_T.
Catalog No. 50-812-683 Supplier New England Biolabs, Inc. Supplier No. R0655S
A restriction endonuclease that recognizes the sequence A^CATG_T.