Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PCK2 Protein, Human (sf9, His-GST) | 94.3% | 10 UG
SDP

Catalog No. 5000400223
Encompass_Preferred

Phosphoenolpyruvate carboxykinase 2 (PCK2) is an enzyme crucial for glucose metabolism, catalyzing the rate-limiting step in converting oxaloacetate (OAA) to phosphoenolpyruvate (PEP). This metabolic pathway is vital for glucose synthesis from lactate and other citric acid cycle precursors. PCK2's activity supports gluconeogenesis, maintaining glucose homeostasis during fasting or high energy demand. By converting OAA to PEP, PCK2 facilitates glucose generation, essential for providing energy to tissues like the brain and red blood cells, thus highlighting its importance in cellular energy balance.

  • Supplied as a 0.22 μm filtered solution
  • Endotoxin level less than 1 EU/μg
  • Stable for 1 year at -80°C, 3 months at -20°C after opening
  • Ships with dry ice
  • Purity 94.3% by reducing SDS-PAGE

Catalog No. 50-004-00223 Supplier Medchemexpress LLC Supplier No. HYP7595710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphoenolpyruvate carboxykinase 2 (PCK2) is an enzyme crucial for glucose metabolism, catalyzing the rate-limiting step in converting oxaloacetate (OAA) to phosphoenolpyruvate (PEP). This metabolic pathway is vital for glucose synthesis from lactate and other citric acid cycle precursors. PCK2's activity supports gluconeogenesis, maintaining glucose homeostasis during fasting or high energy demand. By converting OAA to PEP, PCK2 facilitates glucose generation, essential for providing energy to tissues like the brain and red blood cells, thus highlighting its importance in cellular energy balance.

  • Supplied as a 0.22 μm filtered solution
  • Endotoxin level less than 1 EU/μg
  • Stable for 1 year at -80°C, 3 months at -20°C after opening
  • Ships with dry ice
  • Purity 94.3% by reducing SDS-PAGE

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.