Learn More
Medchemexpress LLC HY-I0660 5g Medchemexpress, PCL 016 CAS:98-98-6 Purity:>98%
SDP
Catalog No. 501933262
Encompass_Preferred
Medchemexpress, HY-I0660 5g PCL 016 CAS:98-98-6 PCL 016 is a topical antiviral agent, which inhibits adenovirus replication in rabbit. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3262 Supplier Medchemexpress LLC Supplier No. HYI06605G
Medchemexpress, HY-I0660 5g PCL 016 CAS:98-98-6 PCL 016 is a topical antiviral agent, which inhibits adenovirus replication in rabbit. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.