Learn More
NANOSOFT POLYMERS PCL-NH2 MW 5000 Size 1g
Catalog No. NC3666408
Encompass
poly(ε-caprolactone) capped with amine (PCL-NH2), is a linear heterofunctional polyester with hydroxyl on one end and NH2 group on the other.PCL are a family of biodegradable and bioactive thermoplastic aliphatic polyester. It is used to fabricate resorbable medical devices that degrade over months in physiological conditions. Due to their history, polylactides are one of the easiest and most affordable biodegradable polymers for medical devices.PCL-NH2can be used to prepare nanoparticles, which can be further modified by NH2-reactive entities.
Catalog No. NC3666408 Supplier NANOSOFT POLYMERS Supplier No. 2759350001G
poly(ε-caprolactone) capped with amine (PCL-NH2), is a linear heterofunctional polyester with hydroxyl on one end and NH2 group on the other.PCL are a family of biodegradable and bioactive thermoplastic aliphatic polyester. It is used to fabricate resorbable medical devices that degrade over months in physiological conditions. Due to their history, polylactides are one of the easiest and most affordable biodegradable polymers for medical devices.PCL-NH2can be used to prepare nanoparticles, which can be further modified by NH2-reactive entities.