Learn More
BEZWADA BIOMEDICAL LLC Poly (caprolactone-co-glycolide) caprolactone:glycolide 85:15, viscosity 0.50 to 1.0 dl/g, 5.0 GM
SDP
Catalog No. 501904457
Encompass_Preferred
The presence of caprolactone repeat units helps in reducing the hardness and toughness of pure polyglycolide polymer and helps in decreasing the hydrolytic degradation rates of polyglycolide homopolymer due to the presence of extended alkyl chain/hydrophobicity of caprolactone.The polymers are used in medical device, drug delivery and 3D printing.
Catalog No. 50-190-4457 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 1281
The presence of caprolactone repeat units helps in reducing the hardness and toughness of pure polyglycolide polymer and helps in decreasing the hydrolytic degradation rates of polyglycolide homopolymer due to the presence of extended alkyl chain/hydrophobicity of caprolactone.The polymers are used in medical device, drug delivery and 3D printing.