Learn More
BEZWADA BIOMEDICAL LLC Poly(caprolactone-co-L-lactide) caprolactone:L-lactide 85:15, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904460
Encompass_Preferred
The presence of lactide repeat units in the copolymer reduces the degree of close packing between caprolactone repeat units leaving the polymer with nice elastic properties while maintaining a relatively slow degradation profile. This allows PCL-PLA copolymers to be used in drug delivery (liquid polymers) and medical device coatings.
Catalog No. 50-190-4460 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 1381
The presence of lactide repeat units in the copolymer reduces the degree of close packing between caprolactone repeat units leaving the polymer with nice elastic properties while maintaining a relatively slow degradation profile. This allows PCL-PLA copolymers to be used in drug delivery (liquid polymers) and medical device coatings.