Learn More
New England Biolabs, Inc. pCLIPf Vector – 20 µg
SDP
Catalog No. 504433
Encompass
pCLIPf Vector is a mammalian expression plasmid intended for the cloning and stable or transient expression of CLIP-tag protein fusions in mammalian cells. This plasmid encodes CLIPf, a CLIP-tag protein, which is expressed under control of the CMV promoter.
Catalog No. 50-443-3 Supplier New England Biolabs, Inc. Supplier No. N9215S
pCLIPf Vector is a mammalian expression plasmid intended for the cloning and stable or transient expression of CLIP-tag protein fusions in mammalian cells. This plasmid encodes CLIPf, a CLIP-tag protein, which is expressed under control of the CMV promoter.