Learn More
TARGETMOL CHEMICALS INC PCLX-001 5MG
SDP
Catalog No. 502579403
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PCLX-001 is a small-molecule compound that acts as an orally active inhibitor of N-myristoyltransferase (NMT) specifically targeting NMT1 and NMT2 with IC50 values of 5 nM and 8 nM respectively. This compound demonstrates anti-tumor effects and effectively inhibits the early signaling of B-cell receptor (BCR). Consequently PCLX-001 is a valuable tool for researching B-cell malignancies [1] [2]. purity: 98%
Catalog No. 50-257-9403 Supplier TARGETMOL CHEMICALS INC Supplier No. T637885MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PCLX-001 is a small-molecule compound that acts as an orally active inhibitor of N-myristoyltransferase (NMT) specifically targeting NMT1 and NMT2 with IC50 values of 5 nM and 8 nM respectively. This compound demonstrates anti-tumor effects and effectively inhibits the early signaling of B-cell receptor (BCR). Consequently PCLX-001 is a valuable tool for researching B-cell malignancies [1] [2]. purity: 98%