Learn More
Aviva Systems Biology Corp PCMT1 Antibody - middle region (ARP48668_T100) | 100uL
SDP
Catalog No. 5001459882
Encompass_Preferred
This is a rabbit polyclonal antibody against PCMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCMT1. Alternate names: PIMTApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59882 Supplier Aviva Systems Biology Corp Supplier No. ARP48668_T100
This is a rabbit polyclonal antibody against PCMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCMT1. Alternate names: PIMTApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish