Learn More
System Biosciences LLC pCMV-CRE, Cre Recombinase Expression PlasmidCre Recombinase Expression Plasmid (pCMV-CRE) = 10ug
SDP
Catalog No. NC1787492
Encompass_Preferred
Transiently express Cre recombinase to remove selection cassettes from your genome engineering projects—use with HR Targeting Vectors that contain LoxP sites. With the Cre-LoxP site-specific recombination system used in most of our HR Targeting Vectors, you can get rid of extra sequences, such as selection cassettes, by transiently expressing the Cre recombinase from the pCMV-CRE Cre Recombinase Expression Plasmid.
Catalog No. NC1787492 Supplier System Biosciences LLC Supplier No. CRE100A1
Transiently express Cre recombinase to remove selection cassettes from your genome engineering projects—use with HR Targeting Vectors that contain LoxP sites. With the Cre-LoxP site-specific recombination system used in most of our HR Targeting Vectors, you can get rid of extra sequences, such as selection cassettes, by transiently expressing the Cre recombinase from the pCMV-CRE Cre Recombinase Expression Plasmid.