Learn More
Cell Biolabs Inc pCMV-Gag-Pol Retroviral Vector, 10 µg
Catalog No. NC9889451
Encompass_Preferred
pCMV-Gag-Pol Retroviral Vector contains both gag and pol genes from MMLV-based retrovirus. This vector may be co-transfected with other retroviral constructs to make recombinant retroviral particles.
Catalog No. NC9889451 Supplier Cell Biolabs Inc Supplier No. RV111
pCMV-Gag-Pol Retroviral Vector contains both gag and pol genes from MMLV-based retrovirus. This vector may be co-transfected with other retroviral constructs to make recombinant retroviral particles.