Learn More
Eurofins Discoverx pCMV- ProLink 2 Vector, 1 vial, 10 µg. (93-0171)
Catalog No. 501999876
Encompass
The pCMV- ProLink 2 Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLink 2 (PK 2) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA, making it ideal for use with weaker protein:protein interactions. Provided as a vial of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/pcmv-2-vector-93-0171.
Catalog No. 50-199-9876 Supplier Eurofins Discoverx Supplier No. 930171
The pCMV- ProLink 2 Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLink 2 (PK 2) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA, making it ideal for use with weaker protein:protein interactions. Provided as a vial of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/pcmv-2-vector-93-0171.