Learn More
Eurofins Discoverx pCMV-ProLabel N Vector, 1 vial, 10 µg. (93-0009)
Catalog No. 501999872
Encompass
The pCMV-ProLabel N Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLabel (ProLabel) tag at the N-terminus. The ProLabel tag is a version of ED used for EFC assays (in combination with EA) where ED and EA are contained in separate cellular compartments. Provided as a vial of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/pcmv-n-vector-93-0009.
Catalog No. 50-199-9872 Supplier Eurofins Discoverx Supplier No. 930009
The pCMV-ProLabel N Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLabel (ProLabel) tag at the N-terminus. The ProLabel tag is a version of ED used for EFC assays (in combination with EA) where ED and EA are contained in separate cellular compartments. Provided as a vial of 10 μg vector in 20 μL liquid buffer. For detailed product information, User Manual and datasheet, visit www.discoverx.com/products/reagent/vector/pcmv-n-vector-93-0009.