Learn More
OriGene PCMV6AC-IRES-GFP-PURO,MAM,10UG
SDP
Catalog No. NC2151636
Encompass_Preferred
pCMV6-AC-IRES-GFP-Puro, mammalian vector with IRES-GFP and puro selection, 10ug
This item is not returnable. View return policy
Catalog No. NC2151636 Supplier OriGene Supplier No. PS100059
pCMV6-AC-IRES-GFP-Puro, mammalian vector with IRES-GFP and puro selection, 10ug