Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5111MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM350. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Tonsil or reactive lymph node. Cellular Staining: Predominantly nuclear, some cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Rat PCNA/Protein A fusion protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 36kDa.Chromosome: 20p12.3. Gene Symbol: PCNA. Gene ID: 5111. Ensemble: ENSG00000132646. Uniprot: NP_002583.
Catalog No. 50-188-5038