Learn More
Aviva Systems Biology Corp PCNP Antibody - middle region (ARP57393_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57393_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCNP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCNP. Alternate names: DKFZp781I24156ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62128