Learn More
TARGETMOL CHEMICALS INC PCO 400 5MG
SDP
Catalog No. 502579025
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PCO 400 is an analog of comakalim that acts as a selective and potent ATP-sensitive K+ channel opener. purity: 98%
Catalog No. 50-257-9025 Supplier TARGETMOL CHEMICALS INC Supplier No. T906045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PCO 400 is an analog of comakalim that acts as a selective and potent ATP-sensitive K+ channel opener. purity: 98%