Learn More
Aviva Systems Biology Corp PCOLCE Antibody - middle region (ARP56378_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56378_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCOLCE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCOLCE. Alternate names: PCPE, PCPE1, PCPE-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61834