Learn More
Creative Biogene PCOMB3XSS VECTOR
SDP
Supplier:  Creative Biogene VPT4013
Encompass
VPT4013 is the modified version of pComb3X. The "SS" refers to the double stuffer, a 1200bp stuffer in the Fab light chain cloning region bounded by SacI and XbaI restriction sites and a 300bp stuffer in Fab heavy chain cloning region bound by XhoI and SpeI restriction sites. Also, the 1600bp double stuffer (both stuffer plus the leader sequence between the Fab light chain and heavy chain cloning regions) can be removed by SfiI digest so that non-Fab genes of interest can also by cloned.
Catalog No. NC1909615