Learn More
Pion Inc PCON GIT-0 LIPID PACK OF 5
Supplier:  Pion Inc 110669
Encompass
Lipid mixture for use in double-sink assays. Contents are frozen under nitrogen. Quantity: 5 ampoules; volume: 1.2 mL per ampoule.
Catalog No. NC0875025