Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 329738
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-112-9997