Learn More
Apexbio Technology LLC pCp-Desthiobiotin 20uL(1 mM)
SDP
Supplier:  Apexbio Technology LLC B88240.02
Encompass_Preferred
pCp-Desthiobiotin (CAS nan) is a chemically modified nucleotide analog intended for labeling RNA or miRNA at the 3 -end It is designed to enable covalent attachment of desthiobiotin to the 3 -hydroxyl terminus of RNA or miRNA using T4 RNA ligase thereby facilitating subsequent affinity-based probe detection pCp-Desthiobiotin exerts its activity through covalent modification of RNA with probe detection relying on interaction between the desthiobiotin moiety and streptavidin conjugates such as horseradish peroxidase (HRP) alkaline phosphatase (AP) or fluorescent labels Based on these properties pCp-Desthiobiotin is widely applicable to hybridization assays including microarray-based RNA expression profiling and electrophoretic mobility shift assays (EMSAs) supporting sensitive analyses in biomedical research
Catalog No. 50-000-89461