Learn More
Aviva Systems Biology Corp PCP2 Antibody - N-terminal region (ARP53294_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53294_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCP2. It was validated on Western Blot. . Alternate names: DKFZp686M0187, FLJ27345, GPSM4, L7, MGC41903ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-823