Learn More
Apexbio Technology LLC PCPA methyl ester hydrochloride 14173-40-1 1g
SDP
Catalog No. 5000092849
Encompass_Preferred
PCPA methyl ester hydrochloride (CAS 14173-40-1) is a lipophilic reversible inhibitor of tryptophan hydroxylase the rate-limiting enzyme responsible for converting tryptophan to serotonin (5-HT) By inhibiting this enzymatic step the compound effectively reduces central serotonin synthesis following blood-brain barrier penetration It is widely utilized in experimental models to investigate serotonergic pathway function serotonin depletion states and their implications in neurobehavioral and physiological processes
Catalog No. 50-000-92849 Supplier Apexbio Technology LLC Supplier No. A47441000
PCPA methyl ester hydrochloride (CAS 14173-40-1) is a lipophilic reversible inhibitor of tryptophan hydroxylase the rate-limiting enzyme responsible for converting tryptophan to serotonin (5-HT) By inhibiting this enzymatic step the compound effectively reduces central serotonin synthesis following blood-brain barrier penetration It is widely utilized in experimental models to investigate serotonergic pathway function serotonin depletion states and their implications in neurobehavioral and physiological processes