Learn More
Roboz Surgical PCPS 4IN 1X2 STR
SDP
Catalog No. NC9827285
Encompass
Micro Dissecting Forceps; 1X2 Teeth, Straight; Extra Delicate; 0.5mm Tip Width; 4 in. Length
Catalog No. NC9827285 Supplier Roboz Surgical Supplier No. RS5152
Micro Dissecting Forceps; 1X2 Teeth, Straight; Extra Delicate; 0.5mm Tip Width; 4 in. Length