Learn More
Sigma Aldrich Fine Chemicals Biosciences PCR 100 bp Low Ladder for
Catalog No. 502900261
Encompass
Sigmas PCR 100 bp Low Marker contains 10 dsDNA fragments from 100 - 1 000 bases evenly spaced in 100 bp repeats. Two ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.
Catalog No. 50-290-0261 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P14731VL
Sigmas PCR 100 bp Low Marker contains 10 dsDNA fragments from 100 - 1 000 bases evenly spaced in 100 bp repeats. Two ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose gel.